Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dhrs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026819.3 | NP_081095.2 | 6 | 879 | ||
GenBank mRNA | AK003958.1 | - | 6 | 769 | ||
AK146317.1 | - | 6 | 826 | |||
AK159964.1 | - | 6 | 801 | |||
AK168115.1 | - | 6 | 826 | |||
AK168147.1 | - | 6 | 826 | |||
AK180428.1 | - | 1 | 51 | |||
BC003930.1 | - | 6 | 810 | 3485584 | ||
BC023825.1 | - | 5 | 1920 |