Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dus1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026824.4 | NP_081100.2 | 13, 14 | 1559 | ||
GenBank mRNA | AK004041.1 | - | 12, 13 | 1082 | ||
AK037538.1 | - | 11, 12 | 2696 | |||
AK049333.1 | - | 430 | ||||
AK088243.1 | - | 13, 14 | 1560 | |||
AK147967.1 | - | 13, 14 | 1759 | |||
AK148307.1 | - | 13, 14 | 1759 | |||
AK179747.1 | - | 1, 2 | 56 | |||
BC019480.1 | - | 11, 12 | 1043 | 4037583 |