Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285812.1 | NP_001272741.1 | 3 | 921 | ||
NM_001285814.1 | NP_001272743.1 | 4 | 986 | |||
NM_001285815.1 | NP_001272744.1 | 4 | 1004 | |||
NM_001285816.1 | NP_001272745.1 | 2 | 784 | |||
NM_026837.3 | NP_081113.1 | 3 | 900 | |||
NM_029589.1 | NP_083865.1 | 4 | 822 | |||
GenBank mRNA | AK004164.1 | - | 3 | 863 | ||
AK005104.1 | - | 3 | 807 | |||
AK005885.1 | - | 4 | 826 | |||
BC019937.1 | - | 3 | 826 | 4023311 | ||
BC020121.1 | - | 3 | 830 | 3991677 | ||
BC039805.1 | - | 2 | 1205 |