Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dcaf12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026893.3 | NP_081169.2 | 9 | 3143 | ||
GenBank mRNA | AK087517.1 | - | 9 | 3027 | ||
AK090028.1 | - | 9 | 3094 | |||
AK143635.1 | - | 9 | 3097 | |||
AK173289.1 | - | 9 | 3003 | |||
BC023852.1 | - | 9 | 3079 | 5346910 |