Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Haghl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271433.1 | NP_001258362.1 | 9 | 1269 | ||
NM_001271434.1 | NP_001258363.1 | 9 | 1271 | |||
NM_001271436.1 | NP_001258365.1 | 8 | 1083 | |||
NM_001271437.1 | NP_001258366.1 | 8 | 1078 | |||
NM_026897.3 | NP_081173.1 | 8 | 1254 | |||
GenBank mRNA | AK005274.1 | - | 8 | 1250 | ||
AK012748.1 | - | 9 | 1268 | |||
AK021220.1 | - | 8 | 1085 | |||
AK032999.1 | - | 5 | 1422 | |||
AK034798.1 | - | 9 | 1246 | |||
AK079632.1 | - | 5 | 1742 | |||
AK080454.1 | - | 1 | 2731 | |||
AK154248.1 | - | 8 | 1059 | |||
BC005586.1 | - | 8 | 1417 | |||
BC030466.1 | - | 9 | 1254 | 5253213 | ||
BC083322.1 | - | 9 | 1208 | 6820885 |