Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zdhhc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026917.5 | NP_081193.2 | 7 | 1826 | ||
GenBank mRNA | AK018755.1 | - | 7 | 1794 | ||
AK080541.1 | - | 7 | 1793 | |||
BC005531.1 | - | 8 | 1853 | |||
BC025126.1 | - | 7 | 1812 | 3669723 | ||
BC052464.1 | - | 7 | 1733 | 6834763 |