Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cnot8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026949.3 | NP_081225.1 | 6, 7 | 877 | ||
GenBank mRNA | AK007581.1 | - | 6, 7 | 941 | ||
AK075727.1 | - | 6, 7 | 847 | |||
AK150757.1 | - | 6, 7 | 958 | |||
AK153907.1 | - | 6, 7 | 880 | |||
AK192037.1 | - | 298 | ||||
AK192809.1 | - | 2, 3 | 214 | |||
AK199523.1 | - | 2, 3 | 204 | |||
AY515608.1 | - | 5, 6 | 843 | |||
BC004040.1 | - | 6, 7 | 877 | 3496188 |