Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pex11g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026951.2 | NP_081227.1 | 5 | 555 | ||
GenBank mRNA | AK007582.1 | - | 5 | 556 | ||
AK007848.1 | - | 4 | 364 | |||
BC024096.1 | - | 5 | 586 | |||
BC062135.1 | - | 5 | 544 | 6510277 |