Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stx18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289535.1 | NP_001276464.1 | 9 | 879 | ||
NM_001289536.1 | NP_001276465.1 | 8 | 801 | |||
NM_026959.3 | NP_081235.2 | 9 | 879 | |||
GenBank mRNA | AK007700.1 | - | 9 | 877 | ||
AK016910.1 | - | 9 | 821 | |||
AK169273.1 | - | 9 | 823 | |||
AK170165.1 | - | 9 | 875 | |||
BC021362.1 | - | 8 | 791 | 5035350 |