Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kbtbd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164574.1 | NP_001158046.1 | 3 | 1460 | ||
NM_026962.3 | NP_081238.2 | 4 | 1492 | |||
GenBank mRNA | AK034334.1 | - | 4 | 1400 | ||
AK078744.1 | - | 3 | 1372 | |||
AK083497.1 | - | 4 | 1474 | |||
BC027392.1 | - | 4 | 1396 | 4954712 |