Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnajc24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026992.3 | NP_081268.1 | 5 | 984 | ||
GenBank mRNA | AK006521.1 | - | 5 | 985 | ||
AK011571.1 | - | 4 | 559 | |||
AY028460.1 | - | 5 | 666 | |||
BC030072.1 | - | 5 | 692 | 3376358 | ||
BC091774.1 | - | 5 | 550 | 5150845 |