Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snx6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026998.3 | NP_081274.2 | 4 | 374 | ||
GenBank mRNA | AK011640.1 | - | 5 | 376 | ||
AK013158.1 | - | 4 | 375 | |||
AK182704.1 | - | 3 | 313 | |||
BC025911.1 | - | 4 | 361 | |||
BC061028.1 | - | 5 | 415 | 6594585 |