Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gtpbp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027000.4 | NP_081276.2 | 10 | 1072 | ||
GenBank mRNA | AF325354.1 | - | 10 | 1056 | ||
AF348208.1 | - | 10 | 1079 | |||
AK141852.1 | - | 10 | 1082 | |||
AK163372.1 | - | 9 | 892 | |||
AK164330.1 | - | 10 | 1073 | |||
AK166711.1 | - | 10 | 1079 | |||
BC005514.1 | - | 10 | 1053 | |||
BC025431.1 | - | 10 | 1077 | 5007174 |