Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rps27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027015.4 | NP_081291.1 | 3, 4 | 256 | ||
GenBank mRNA | AA036204.1 | - | 3, 4 | 410 | ||
AK014294.1 | - | 3, 4 | 256 | |||
AK084058.1 | - | 3, 4 | 256 | |||
AK088320.1 | - | 3, 4 | 259 | |||
AK166551.1 | - | 3, 4 | 257 | |||
BC037026.1 | - | 3, 4 | 256 | |||
BC048352.1 | - | 3, 4 | 240 | 5008906 | ||
BC055693.1 | - | 3, 4 | 246 | 6408041 |
Gene Symbol: Rps27rt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190258.1 | NP_001177187.1 | 1 | 255 |