Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cmtm2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027022.4 | NP_081298.1 | 4, 5 | 633 | ||
GenBank mRNA | AK005603.1 | - | 4, 5 | 586 | ||
AK161303.1 | - | 4, 5 | 627 | |||
AY241871.1 | - | 4, 5 | 633 | |||
BC049747.1 | - | 4, 5 | 626 | 6772661 |