Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Avpi1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027106.4 | NP_081382.1 | 3 | 544 | ||
GenBank mRNA | AK009243.1 | - | 3 | 545 | ||
AK177091.1 | - | 1 | 36 | |||
BC027646.1 | - | 3 | 578 | 1344413 |