Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LAMB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000228.2 | NP_000219.2 | 5 | 487 | ||
NM_001017402.1 | NP_001017402.1 | 4 | 733 | |||
NM_001127641.1 | NP_001121113.1 | 5 | 461 | |||
XM_005273124.4 | XP_005273181.1 | 5 | 669 | |||
XM_017001272.1 | XP_016856761.1 | 5 | 1089 | |||
GenBank mRNA | AK296851.1 | - | 5 | 437 | ||
AK309524.1 | - | 5 | 437 | |||
AL555685.3 | - | 5 | 487 | |||
AY035783.1 | - | 4 | 343 | |||
BC075838.1 | - | 4 | 518 | 6291096 | ||
BP350980.1 | - | 5 | 503 | |||
D37766.1 | - | 5 | 424 | |||
KU178159.1 | - | 4 | 343 | |||
L25541.1 | - | 5 | 463 |