Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx51 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027156.3 | NP_081432.2 | 11 | 1595 | ||
GenBank mRNA | AK010015.1 | - | 2 | 199 | ||
AK141361.1 | - | 11 | 1576 | |||
AK152599.1 | - | 11 | 1563 | |||
BC023700.1 | - | 8 | 858 | |||
BC060646.1 | - | 11 | 1572 | 6853826 | ||
BC069876.1 | - | 4 | 423 |