Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040111.1 | NP_001035200.1 | 34 | 4956 | ||
NM_001040112.1 | NP_001035201.1 | 31 | 4433 | |||
NM_027180.3 | NP_081456.2 | 30 | 4048 | |||
NM_198096.1 | NP_932764.1 | 21 | 2792 | |||
GenBank mRNA | AK010344.1 | - | 2 | 514 | ||
AK052437.1 | - | 21 | 2789 | |||
AK129213.1 | - | 28 | 3830 | |||
AK137455.1 | - | 34 | 4930 | |||
AK140646.1 | - | 34 | 4905 | |||
AK150132.1 | - | 31 | 4367 | |||
BC042640.1 | - | 24 | 3222 | |||
BC057922.1 | - | 5 | 754 | |||
BC069849.1 | - | 2 | 555 | |||
BC083088.1 | - | 1 | 406 | |||
BC091762.1 | - | 5 | 728 | |||
BC141179.1 | - | 33 | 4826 | 9055791 |