Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tm2d2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027194.3 | NP_081470.2 | 2 | 454 | ||
GenBank mRNA | AK010545.1 | - | 2 | 386 | ||
AK045768.1 | - | 2 | 413 | |||
AK080854.1 | - | 2 | 382 | |||
AK083654.1 | - | 2 | 375 | |||
AK085975.1 | - | 2 | 406 | |||
AK086015.1 | - | 2 | 392 | |||
AK086059.1 | - | 2 | 408 | |||
AK088246.1 | - | 2 | 406 | |||
AK149407.1 | - | 2 | 383 | |||
AK160039.1 | - | 2 | 391 | |||
AK166012.1 | - | 2 | 407 | |||
AK206565.1 | - | 1 | 33 | |||
BC026789.1 | - | 2 | 381 | 4188168 |