Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LEP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000230.2 | NP_000221.1 | 3 | 596 | ||
XM_005250340.4 | XP_005250397.1 | 3 | 670 | |||
GenBank mRNA | AF008123.1 | - | 2 | 556 | ||
BC060830.1 | - | 3 | 594 | 30333193 | ||
BC069323.1 | - | 2 | 590 | 7262097 | ||
BC069452.1 | - | 2 | 590 | 7262109 | ||
BC069527.1 | - | 2 | 590 | 7262121 | ||
U43653.1 | - | 2 | 595 |