Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bdh2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172055.1 | NP_001165526.1 | 3 | 325 | ||
NM_027208.2 | NP_081484.1 | 3 | 252 | |||
GenBank mRNA | AK208132.1 | - | 1 | 31 | ||
BC021461.1 | - | 3 | 330 | |||
BC036998.1 | - | 3 | 192 | 5041996 |