Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ms4a6b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027209.3 | NP_081485.2 | 5 | 599 | ||
GenBank mRNA | AF237909.1 | - | 5 | 599 | ||
AK007613.1 | - | 530 | ||||
AK075671.1 | - | 5 | 576 | |||
AK172345.1 | - | 673 | ||||
BC027425.1 | - | 5 | 576 | 3984299 |