Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mzb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027222.3 | NP_081498.2 | 2 | 243 | ||
GenBank mRNA | AK008016.1 | - | 2 | 244 | ||
AK088094.1 | - | 2 | 263 | |||
BC030674.1 | - | 1 | 25 | |||
BC064044.1 | - | 2 | 230 | 875322 | ||
GQ477351.1 | - | 2 | 209 |