Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Coa7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027250.4 | NP_081526.1 | 3 | 1108 | ||
GenBank mRNA | AK044999.1 | - | 3 | 1109 | ||
AK081388.1 | - | 3 | 1121 | |||
AK168902.1 | - | 1048 |