Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il17rd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134437.3 | NP_602319.1 | 13 | 7728 | ||
GenBank mRNA | AK012666.1 | - | 1 | 534 | ||
BC065776.1 | - | 1 | 408 | |||
BC106187.1 | - | 1 | 496 |