Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdc16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027276.2 | NP_081552.2 | 13 | 1346 | ||
GenBank mRNA | AK013213.1 | - | 2 | 280 | ||
AK147938.1 | - | 13 | 1361 | |||
AK167965.1 | - | 13 | 1361 | |||
AK168978.1 | - | 13 | 1333 | |||
AK201828.1 | - | 1 | 65 | |||
BC026606.1 | - | 13 | 1346 | 4976424 |