Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpp25l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027278.3 | NP_081554.2 | 2 | 671 | ||
GenBank mRNA | AJ293897.1 | - | 2 | 666 | ||
AK013222.1 | - | 2 | 660 | |||
BC047068.1 | - | 2 | 663 | 5009034 |