Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Manba | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027288.3 | NP_081564.3 | 1, 2 | 255 | ||
GenBank mRNA | AF306557.1 | - | 1, 2 | 259 | ||
AK036505.1 | - | 1, 2 | 253 | |||
AK047057.1 | - | 1, 2 | 248 | |||
AK142165.1 | - | 1, 2 | 220 | |||
AK154172.1 | - | 1, 2 | 233 | |||
AK166022.1 | - | 1, 2 | 237 | |||
AK166340.1 | - | 1, 2 | 253 | |||
AK170290.1 | - | 1, 2 | 252 | |||
AK171416.1 | - | 1, 2 | 242 | |||
BC031409.1 | - | 1, 2 | 198 | 4952710 |