Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dennd6b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027081.3 | NP_081357.2 | 13 | 1176 | ||
GenBank mRNA | AK006427.1 | - | 4 | 313 | ||
AK084555.1 | - | 12 | 1267 | |||
AK154949.1 | - | 11 | 1431 | |||
AK180459.1 | - | 3 | 203 | |||
BC017544.1 | - | 7 | 706 | |||
BC030395.1 | - | 13 | 1430 | |||
BC058958.1 | - | 9 | 683 | |||
BC139003.1 | - | 13 | 1150 | 8862025 | ||
BQ444357.1 | - | 9 | 682 |