Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040111.1 | NP_001035200.1 | 34 | 5016 | ||
NM_001040112.1 | NP_001035201.1 | 31 | 4493 | |||
NM_027180.3 | NP_081456.2 | 30 | 4108 | |||
NM_198096.1 | NP_932764.1 | 21 | 2852 | |||
GenBank mRNA | AK010344.1 | - | 2 | 574 | ||
AK052437.1 | - | 21 | 2849 | |||
AK137455.1 | - | 34 | 4990 | |||
AK140646.1 | - | 34 | 4965 |