Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rasip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028544.1 | NP_082820.1 | 10 | 2437 | ||
GenBank mRNA | AK011557.2 | - | 4 | 520 | ||
AK033276.1 | - | 8 | 1725 | |||
AK156602.1 | - | 10 | 2449 | |||
BC028444.1 | - | 509 | ||||
BC072584.1 | - | 7 | 1391 | |||
BC127059.1 | - | 8 | 1363 | 40109852 | ||
BC150766.1 | - | 10 | 2437 | 9087677 |