Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptcd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027275.3 | NP_081551.2 | 20 | 1640 | ||
GenBank mRNA | AK013131.1 | - | 6 | 419 | ||
AK146005.1 | - | 20 | 1629 | |||
AK146172.1 | - | 20 | 1641 | |||
AK172480.1 | - | 21 | 1705 | |||
BC026759.1 | - | 4 | 295 | |||
BC055844.1 | - | 6 | 467 | |||
BC060687.1 | - | 11 | 861 | |||
BC084595.1 | - | 11 | 890 | |||
BC115434.2 | - | 20 | 1636 | 40094203 |