Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nkd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163660.1 | NP_001157132.1 | 4, 5 | 423 | ||
NM_027280.3 | NP_081556.3 | 6, 7 | 633 | |||
GenBank mRNA | AF343352.1 | - | 6, 7 | 470 | ||
AF358134.1 | - | 6, 7 | 549 | |||
AK076164.1 | - | 6, 7 | 624 | |||
AK082367.1 | - | 6, 7 | 622 | |||
AK166564.1 | - | 6, 7 | 636 | |||
BC034838.1 | - | 6, 7 | 607 | 1429943 | ||
DV662736.1 | - | 4, 5 | 403 |