Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gemin7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027189.2 | NP_081465.1 | 2 | 630 | ||
GenBank mRNA | AK010303.1 | - | 2 | 632 | ||
BC028527.1 | - | 2 | 626 | 1362579 |