Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ms4a6b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027209.3 | NP_081485.2 | 7 | 1163 | ||
GenBank mRNA | AF237909.1 | - | 7 | 1163 | ||
AK007613.1 | - | 1094 | ||||
AK075671.1 | - | 7 | 1140 | |||
AK149773.1 | - | 197 | ||||
AK172345.1 | - | 2911 | ||||
BC027425.1 | - | 7 | 1140 | 3984299 |
Gene Symbol: Ms4a6c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028595.4 | NP_082871.2 | 7 | 1527 | ||
GenBank mRNA | AK150397.1 | - | 7 | 1423 | ||
AK152258.1 | - | 7 | 1425 | |||
BY691481.1 | - | 1 | 81 |