Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptcd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027275.3 | NP_081551.2 | 24 | 2422 | ||
GenBank mRNA | AK013131.1 | - | 10 | 1201 | ||
AK146005.1 | - | 24 | 2411 | |||
AK146172.1 | - | 24 | 2429 | |||
AK172480.1 | - | 25 | 2497 | |||
BC026759.1 | - | 8 | 1078 | |||
BC055844.1 | - | 9 | 2196 | |||
BC060687.1 | - | 15 | 1643 | |||
BC068263.1 | - | 3 | 1005 | |||
BC084595.1 | - | 15 | 1673 |