Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: March5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164336.1 | NP_001157808.1 | 2 | 434 | ||
NM_001164337.1 | NP_001157809.1 | 2 | 434 | |||
NM_027314.3 | NP_081590.3 | 2 | 434 | |||
GenBank mRNA | AK007511.1 | - | 2 | 270 | ||
AK009232.1 | - | 2 | 420 | |||
AK009364.1 | - | 2 | 433 | |||
AK021403.1 | - | 2 | 404 | |||
BC064752.1 | - | 2 | 158 | 30049608 | ||
BC107214.1 | - | 2 | 145 | |||
BC107215.1 | - | 2 | 146 | 40051628 | ||
BC132455.1 | - | 2 | 198 | 40130732 | ||
BC132457.1 | - | 2 | 197 | 40130734 | ||
BY070559.1 | - | 2 | 327 |