Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sfxn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027324.5 | NP_081600.1 | 2 | 250 | ||
GenBank mRNA | AF325260.1 | - | 2 | 228 | ||
AK012650.1 | - | 2 | 249 | |||
AK151110.1 | - | 2 | 196 | |||
AK179717.1 | - | 1 | 35 | |||
BC005743.1 | - | 2 | 207 | 3599636 |