Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mllt3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286158.1 | NP_001273087.1 | 2, 3 | 189 | ||
NM_027326.3 | NP_081602.3 | 6, 7 | 1516 | |||
NM_029931.3 | NP_084207.2 | 2, 3 | 189 | |||
GenBank mRNA | AF333960.1 | - | 5, 6 | 1177 | ||
AK008707.1 | - | 2, 3 | 151 | |||
AK019458.1 | - | 2, 3 | 181 | |||
AK136066.1 | - | 2, 3 | 749 | |||
AK147009.1 | - | 2, 3 | 103 | |||
BC021420.1 | - | 7, 8 | 1355 | 5012671 | ||
BU614509.1 | - | 2, 3 | 317 | |||
BY713607.1 | - | 2, 3 | 177 | |||
DV051568.1 | - | 199 |