Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTCH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000264.3 | NP_000255.2 | 6 | 1048 | ||
NM_001083602.1 | NP_001077071.1 | 6 | 1156 | |||
NM_001083603.1 | NP_001077072.1 | 6 | 1002 | |||
NM_001083604.1 | NP_001077073.1 | 6 | 766 | |||
NM_001083605.1 | NP_001077074.1 | 6 | 749 | |||
NM_001083606.1 | NP_001077075.1 | 6 | 848 | |||
NM_001083607.1 | NP_001077076.1 | 6 | 922 | |||
GenBank mRNA | AB189436.1 | - | 6 | 1156 | ||
AB189437.1 | - | 6 | 1002 | |||
AB189438.1 | - | 6 | 768 | |||
AB189439.1 | - | 6 | 751 | |||
AB189440.1 | - | 6 | 921 | |||
AB209495.1 | - | 3 | 497 | |||
DB093644.1 | - | 2 | 373 | |||
U43148.1 | - | 6 | 848 | |||
U59464.1 | - | 6 | 860 |