Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spdl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027411.2 | NP_081687.2 | 4, 5 | 704 | ||
GenBank mRNA | AK011138.1 | - | 4, 5 | 676 | ||
AK012927.1 | - | 4, 5 | 705 | |||
AK139553.1 | - | 4, 5 | 580 | |||
BC006674.1 | - | 4, 5 | 687 | 3499487 | ||
BC025900.1 | - | 1, 2 | 622 | |||
BY712013.1 | - | 4, 5 | 705 |