Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Calml3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027416.3 | NP_081692.1 | 1 | 274 | ||
GenBank mRNA | AK010118.1 | - | 1 | 272 | ||
BC005457.1 | - | 1 | 269 | 3493620 | ||
EL960451.1 | - | 1 | 274 |