Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr77 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027432.3 | NP_081708.1 | 2 | 371 | ||
GenBank mRNA | AK012014.1 | - | 2 | 362 | ||
AK048488.1 | - | 2 | 394 | |||
AK050391.1 | - | 2 | 371 | |||
AK078856.1 | - | 3 | 330 | |||
AK131608.1 | - | 1 | 282 | |||
AK161812.1 | - | 2 | 346 | |||
AK163839.1 | - | 366 | ||||
AK168006.1 | - | 2 | 373 | |||
AK168752.1 | - | 2 | 393 | |||
AK169128.1 | - | 2 | 372 | |||
AK191398.1 | - | 1 | 89 | |||
BC005755.1 | - | 2 | 326 | 2654151 |