Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp398 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027477.3 | NP_081753.1 | 3 | 763 | ||
NM_173034.3 | NP_766622.2 | 2 | 367 | |||
GenBank mRNA | AK078250.1 | - | 2 | 334 | ||
AK081169.1 | - | 3 | 728 | |||
AK089190.1 | - | 3 | 730 | |||
AK155461.1 | - | 3 | 730 | |||
BC112382.1 | - | 3 | 689 | 40059618 |