Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Medag | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027519.3 | NP_081795.1 | 2 | 500 | ||
GenBank mRNA | AB055406.1 | - | 2 | 532 | ||
AK018128.1 | - | 2 | 316 | |||
AK036605.1 | - | 2 | 501 | |||
AK042557.1 | - | 2 | 213 | |||
AK047657.1 | - | 2 | 557 | |||
AK053969.1 | - | 2 | 534 | |||
AK086092.1 | - | 2 | 509 | |||
AK138268.1 | - | 2 | 199 | |||
AK160904.1 | - | 2 | 507 | |||
AK161526.1 | - | 2 | 509 | |||
AK162725.1 | - | 2 | 501 | |||
AK163617.1 | - | 2 | 199 | |||
AK165529.1 | - | 2 | 297 | |||
BC026658.1 | - | 2 | 263 | |||
BC116246.1 | - | 2 | 422 | 40093530 | ||
BC116247.1 | - | 2 | 420 | 40093536 |