Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Meaf6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290701.1 | NP_001277630.1 | 3 | 284 | ||
NM_027310.4 | NP_081586.1 | 3 | 284 | |||
GenBank mRNA | AK009177.1 | - | 3 | 272 | ||
AK012867.1 | - | 3 | 256 | |||
BC061024.1 | - | 3 | 254 | |||
BC108404.1 | - | 3 | 255 | 30135798 |