Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nars | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142950.1 | NP_001136422.1 | 2, 3 | 233 | ||
NM_027350.3 | NP_081626.2 | 2, 3 | 230 | |||
GenBank mRNA | AK077699.1 | - | 2, 3 | 211 | ||
AK083730.1 | - | 2, 3 | 190 | |||
AK157244.1 | - | 2, 3 | 192 | |||
AK172657.1 | - | 2, 3 | 192 | |||
AK191424.1 | - | 1, 2 | 51 | |||
BC052849.1 | - | 2, 3 | 191 | 30057822 | ||
BY117114.1 | - | 2, 3 | 234 |