Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aldh1l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027406.1 | NP_081682.1 | 4 | 542 | ||
GenBank mRNA | BC022112.1 | - | 4 | 542 | ||
BC024055.1 | - | 4 | 499 | 5098789 | ||
BC025939.1 | - | 4 | 524 | 5099360 | ||
BC028817.1 | - | 4 | 489 | 5101815 | ||
BC030722.1 | - | 4 | 526 | 5098671 | ||
BC030723.1 | - | 4 | 458 | 5100007 | ||
BC030727.1 | - | 4 | 528 | 5099026 | ||
BC030730.1 | - | 4 | 542 | 5101835 |