Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 2810408A11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027419.3 | NP_081695.2 | 1 | 570 | ||
GenBank mRNA | AK013042.1 | - | 1 | 571 | ||
AK015537.1 | - | 1 | 502 | |||
AK051029.1 | - | 1 | 542 | |||
AK160144.1 | - | 1 | 479 | |||
AK171674.1 | - | 1 | 501 | |||
BC051546.1 | - | 1 | 531 | 5255109 | ||
BC055710.1 | - | 1 | 559 | |||
BC069874.1 | - | 1 | 570 | 4216824 |